4-Chlorobutyl acetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22582-14
B22582-22
B22582-36
CAAAB22582-14EA
64.15
CAD
CAAAB22582-14
CAAAB22582-22
CAAAB22582-36
4-Chlorobutyl acetate 98%
4-Chlorobutyl acetate
Purity: 98%
CAS No.: 6962-92-1
Molecular Formula: CH3CO2(CH2)4Cl
Formula Weight: 150.61
Flash Point: 64°(147°F)
Refractive Index: 1.4350
Boiling Point: 81-83°/11mm
Beilstein Reference: 1749681
MDL No.: MFCD00001013
Formula:
C₆H₁₁ClO₂ Boiling Pt: 81…83 °C/11 mm Density: 1.076 Flash Pt: 64 °C(147 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001013 CAS Number: 6962-92-1 EINECS: 230-158-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...