3,4,5-Trifluorobenzyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22578-03
B22578-06
CAAAB22578-03EA
99.86
CAD
CAAAB22578-03
CAAAB22578-06
3,4,5-Trifluorobenzyl bromide 97%
3,4,5-Trifluorobenzyl bromide
Purity: 97%
CAS No.: 220141-72-0
Molecular Formula: C7H4BrF3
Formula Weight: 225.01
Flash Point: 65°(149°F)
Refractive Index: 1.5050
Boiling Point: 44-46°/1mm
Beilstein Reference: 8680985
UN No.: 3265
MDL No.: MFCD00083528
Formula:
C₇H₄BrF₃ Boiling Pt: 44…46 °C/1 mm Density: 1.689 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00083528 CAS Number: 220141-72-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...