3-Fluoro-N-methylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22565-09
B22565-18
CAAAB22565-09EA
143.78
CAD
CAAAB22565-09
CAAAB22565-18
CAAAB22565-30
3-Fluoro-N-methylaniline 97%
3-Fluoro-N-methylaniline
Purity: 97%
CAS No.: 1978-37-6
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 86°(187°F)
Odor: Characteristic
Boiling Point: 94°/19mm
UN No.: 2810
MDL No.: MFCD02683095
Formula:
C₇H₈FN Boiling Pt: 94 °C/19 mm Density: 1.11 Flash Pt: 86 °C (187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02683095 CAS Number: 1978-37-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...