Methyl methoxyacetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methoxyacetic methyl ester
B22528-22
B22528-36
CAAAB22528-22EA
130.35
CAD
CAAAB22528-22
CAAAB22528-36
Methyl methoxyacetate 99%
Methyl methoxyacetate
Purity: 99%
CAS No.: 6290-49-9
Molecular Formula: CH3OCH2CO2CH3
Formula Weight: 104.11
Flash Point: 35°(95°F)
Refractive Index: 1.3960
Boiling Point: 129-130°
Beilstein Reference: 1742944
UN No.: 3272
MDL No.: MFCD00008451
Formula:
C₄H₈O₃ Boiling Pt: 129…130 °C Density: 1.051 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008451 CAS Number: 6290-49-9 EINECS: 228-539-9 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...