(3-Chloropropyl)trimethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloropropyltrimethoxysilane, 3-(Trimethoxysilyl)propylchloride, Trimethoxy(3-chloropropyl)silane

B22517-22 B22517-36
CAAAAB22517-22EA 65.1 CAD
CAAAAB22517-22 CAAAAB22517-36
(3-Chloropropyl)trimethoxysilane 97%
(3-Chloropropyl)trimethoxysilane

Purity: 97% CAS No.: 2530-87-2 Molecular Formula: ClCH2CH2CH2Si(OCH3)3 Formula Weight: 198.72 Flash Point: 78°(172°F) Refractive Index: 1.4200 Odor: Recognizable Melting Point: -50° Boiling Point: 195-197° Beilstein Reference: 969627 MDL No.: MFCD00000997

Formula: ClH₂CCH₂CH₂Si(OCH₃)₃
MW: 198.72 g/mol
Boiling Pt: 195…197 °C
Melting Pt: –50 °C
Density: 1.077
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000997
CAS Number: 2530-87-2
EINECS: 219-787-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR