Zinc bromide, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22510-22
B22510-36
CAAAAB22510-22EA
74.89
CAD
CAAAAB22510-22
CAAAAB22510-36
Zinc bromide, anhydrous 98%
Zinc bromide
Purity: 98%
CAS No.: 7699-45-8
Molecular Formula: ZnBr2
Formula Weight: 225.19
Refractive Index: 1.5452
Melting Point: 394°
Boiling Point: 650°
Merck Reference: 14,10129
UN No.: 3260
MDL No.: MFCD00011294
Formula:
Br₂Zn Boiling Pt: 650 °C Melting Pt: 394 °C Density: 4.201 Storage Temperature: Ambient |
MDL Number:
MFCD00011294 CAS Number: 7699-45-8 EINECS: 231-718-4 UN: 3260 ADR: 8,III Merck Index: 14,10129 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...