Magnesium silicate 60-120 mesh for chromatography

Supplier: Thermo Scientific Chemicals

B22490-0B B22490-30
CAAAB22490-0BEA 568.8 CAD
CAAAB22490-0B CAAAB22490-30
Magnesium silicate 60-120 mesh for chromatography
Magnesium silicate

Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
Notes: Incompatible with strong acids and hydrogen fluoride.

Formula: Mg₂O₄Si
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR