Phosphorus(V) bromide 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phosphorus pentabromide

B22481-14 B22481-22 B22481-36
CAAAB22481-14EA 142.99 CAD
CAAAB22481-14 CAAAB22481-22 CAAAB22481-36
Phosphorus(V) bromide 95%
Phosphorus (V) bromide

Purity: 95% CAS No.: 7789-69-7 Molecular Formula: PBr5 Formula Weight: 430.50 Odor: Acrid Melting Point: ca 105° dec. Boiling Point: 105-107° dec. Beilstein Reference: 3903332 Merck Reference: 14,7350 UN No.: 2691 MDL No.: MFCD00011437

Formula: PBr₅
MW: 430.49
Boiling Pt: 105…107 °C (dec.)
Melting Pt: ∼ 105 °C (dec.)
Density: 3.61
Storage Temperature: Ambient
MDL Number: MFCD00011437
CAS Number: 7789-69-7
EINECS: 232-186-6
UN: 2691
ADR: 8 II
Merck Index: 14,7350

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR