Cesium hydroxide monohydrate 96% CsCO₃ <5%

Supplier: Thermo Scientific Chemicals
Danger

B22480-06 B22480-14 B22480-22 B22480-36
CAAAAB22480-06EA 54.67 CAD
CAAAAB22480-06 CAAAAB22480-14 CAAAAB22480-22 CAAAAB22480-36
Cesium hydroxide monohydrate 96% CsCO₃ <5%
Cesium hydroxide monohydrate

Purity: 96% CAS No.: 35103-79-8 Molecular Formula: CsOH.H2O Formula Weight: 167.93 (149.91anhy) Odor: Odorless Melting Point: 205-206° UN No.: 3262 MDL No.: MFCD00149664

Formula: HCsO·1H₂O
Melting Pt: 205…206 °C
Density: 1.870
Storage Temperature: Ambient
MDL Number: MFCD00149664
CAS Number: 35103-79-8
EINECS: 244-344-1
UN: 2682
ADR: 8,II
Merck Index: 14,2013

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR