Tris(trimethylsilyl)silane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22457-03
B22457-06
B22457-14
CAAAB22457-03EA
59.25
CAD
CAAAB22457-03
CAAAB22457-06
CAAAB22457-14
Tris(trimethylsilyl)silane 97%
Tris(trimethylsilyl)silane
Purity: 97%
CAS No.: 1873-77-4
Molecular Formula: [(CH3)3Si]3SiH
Formula Weight: 248.67
Flash Point: 55°(131°F)
Refractive Index: 1.4900
Boiling Point: 82-84°/12mm
Beilstein Reference: 1923953
Merck Reference: 14,9757
UN No.: 1993
MDL No.: MFCD00077893
Formula:
C₉H₂₈Si₄ Boiling Pt: 82…84 °C/12 mm Density: 0.806 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00077893 CAS Number: 1873-77-4 UN: 1993 ADR: 3,III Merck Index: 14,9757 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...