1-Hexylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22446-09
B22446-88
CAAAB22446-09EA
464.52
CAD
CAAAB22446-09
CAAAB22446-88
1-Hexylboronic acid 97%
1-Hexylboronic acid
Purity: 97%
CAS No.: 16343-08-1
Molecular Formula: CH3(CH2)5B(OH)2
Formula Weight: 130.00
Melting Point: 85-90°
Beilstein Reference: 1736080
Formula:
C₆H₁₅BO₂ Melting Pt: 85…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074641 CAS Number: 16343-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...