Perfluoro-tert-butanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Perfluoro-t-butanol, Nonafluoro-tert-butyl alcohol, 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butyl alcohol

B22442-06 B22442-14
CAAAAB22442-06EA 440.82 CAD
CAAAAB22442-06 CAAAAB22442-14
Perfluoro-tert-butanol 99%
Perfluoro-tert-butanol

Purity: 99% CAS No.: 2378-02-1 Molecular Formula: C4HF9O Formula Weight: 236.04 Flash Point: None Refractive Index: 1.3 00 Odor: Characteristic Melting Point: -17° Boiling Point: 44-45° Beilstein Reference: 1841640 UN No.: 2810 MDL No.: MFCD00042092

Formula: C₄HF₉O
MW: 236.04 g/mol
Boiling Pt: 44…45 °C
Melting Pt: –17 °C
Density: 1.693
Storage Temperature: Ambient
MDL Number: MFCD00042092
CAS Number: 2378-02-1
EINECS: 219-157-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR