(±)-3-Methyl-1-pentanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methylpentan-1-ol, (±)-3-Methylpentan-1-ol, 3-Methyl-1-pentanol
B22356-06
B22356-14
CAAAB22356-06EA
80.26
CAD
CAAAB22356-06
CAAAB22356-14
(±)-3-Methyl-1-pentanol 98%
(±)-3-Methyl-1-pentanol
Purity: 98%
CAS No.: 589-35-5
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 58°(136°F)
Refractive Index: 1.4180
Boiling Point: 151-152°
Beilstein Reference: 1718979
UN No.: 1987
MDL No.: MFCD00002937
Formula:
C₆H₁₄O MW: 102.18 g/mol Boiling Pt: 151…152 °C Melting Pt: -70 Density: 0.823 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002937 CAS Number: 589-35-5 EINECS: 209-644-9 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...