(S)-(-)-1,2-Epoxypropane 99%

Supplier: Thermo Scientific Chemicals
Danger

B22341-06 B22341-14
CAAAB22341-06EA 156.58 CAD
CAAAB22341-06 CAAAB22341-14 CAAAB22341-22
(S)-(-)-1,2-Epoxypropane 99%
(S)-(-)-1,2-Epoxypropane

Purity: 99% CAS No.: 16088-62-3 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -37°(-35°F) Refractive Index: 1.3670 Odor: Ether-like Boiling Point: 34-35° Beilstein Reference: 79765 Merck Reference: 14,7856 Optical Rotation: -14° (neat) UN No.: 1280 MDL No.: MFCD00064312

Formula: C₃H₆O
Boiling Pt: 34…35 °C
Melting Pt: –112 °C
Density: 0.829
Flash Pt: –37 °C (–35 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064312
CAS Number: 16088-62-3
EINECS: 240-241-0
UN: 1280
ADR: 3,I
Merck Index: 14,7856

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR