2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Iodonitrotetrazolium chloride, p-Iodonitrotetrazolium violet, INT, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazol-3-ium chloride, Iodonitrotetrazolium violet, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyltetrazolium chloride

B22301-03 B22301-06 B22301-14
CAAAAB22301-03EA 129.88 CAD
CAAAAB22301-03 CAAAAB22301-06 CAAAAB22301-14
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride 95%
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride

Used for dehydrogenase determination


Purity: 95% CAS No.: 146-68-9 Molecular Formula: C19H13ClIN5O2.xH2O Formula Weight: 505.70(anhy) Melting Point: ca 231° dec. Beilstein Reference: 4093224

Formula: C₁₉H₁₃ClIN₅O₂
MW: 505.7 g/mol
Melting Pt: ∼ 231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011961
CAS Number: 146-68-9
EINECS: 205-676-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR