trans-2-Hexene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22289-09
B22289-88
CAAAAB22289-09EA
276.5
CAD
CAAAAB22289-09
CAAAAB22289-88
trans-2-Hexene 99%
trans-2-Hexene
Purity: 99%
CAS No.: 4050-45-7
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -20°(-4°F)
Refractive Index: 1.3940
Odor: Mild
Melting Point: -99°
Boiling Point: 68-69°
Beilstein Reference: 1719020
UN No.: 3295
MDL No.: MFCD00009473
Formula:
C₆H₁₂ Boiling Pt: 68…69 °C Melting Pt: –99 °C Density: 0.674 Flash Pt: –20 °C (–4 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009473 CAS Number: 4050-45-7 EINECS: 223-752-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...