4,4'-Bis(diethylamino)benzophenone ≥99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone, 4,4-Bis(diethylamino) benzophenone

B22288-14 B22288-22
CAAAAB22288-14EA 83.74 CAD
CAAAAB22288-14 CAAAAB22288-22
4,4'-Bis(diethylamino)benzophenone ≥99%
4,4'-Bis(diethylamino)benzophenone

Purity: 99% CAS No.: 90-93-7 Molecular Formula: C21H28N2O Formula Weight: 324.47 Flash Point: 151°(303°F) Odor: Odorless pH: 4-6 Melting Point: 93-95° Beilstein Reference: 2148611 UN No.: 3077 MDL No.: MFCD00009044

Formula: C₂₁H₂₈N₂O
MW: 324.47 g/mol
Boiling Pt: 420
Melting Pt: 96.5
Flash Pt: 151 °C (303 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009044
CAS Number: 90-93-7
EINECS: 202-025-4
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR