3,5-Dimethoxyphenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22283-09
B22283-18
CAAAAB22283-09EA
140.94
CAD
CAAAAB22283-09
CAAAAB22283-18
CAAAAB22283-30
3,5-Dimethoxyphenol 98%
3,5-Dimethoxyphenol
Purity: 98%
CAS No.: 500-99-2
Molecular Formula: C8H10O3
Formula Weight: 154.16
Flash Point: 78°(173°F)
Melting Point: 45-47°
Boiling Point: 172-175°/17mm
Beilstein Reference: 1239156
MDL No.: MFCD00008388
Formula:
C₈H₁₀O₃ Boiling Pt: 172…175 °C/17 mm Melting Pt: 45…47 °C Flash Pt: 78 °C(173 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008388 CAS Number: 500-99-2 EINECS: 207-917-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...