2,3-Dimethylbutane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22223-14
B22223-22
CAAAAB22223-14EA
238.58
CAD
CAAAAB22223-14
CAAAAB22223-22
2,3-Dimethylbutane 99%
2,3-Dimethylbutane
Purity: 99%
CAS No.: 79-29-8
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -29°(-20°F)
Refractive Index: 1.375
Odor: Mild
Melting Point: -129°
Boiling Point: 57-59°
Beilstein Reference: 1730737
UN No.: 2457
MDL No.: MFCD00008925
Formula:
C₆H₁₄ Boiling Pt: 57…59 °C Melting Pt: –129 °C Density: 0.662 Flash Pt: –29 °C (–20 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008925 CAS Number: 79-29-8 EINECS: 201-193-6 UN: 2457 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...