Diethoxy(diphenyl)silane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22204-18
B22204-30
CAAAAB22204-09EA
37.98
CAD
CAAAAB22204-09
CAAAAB22204-18
CAAAAB22204-30
Diethoxy(diphenyl)silane 98%
Diethoxy(diphenyl)silane
Purity: 98%
CAS No.: 2553-19-7
Molecular Formula: C16H20O2Si
Formula Weight: 272.42
Flash Point: 175°(347°F)
Refractive Index: 1.5255
Boiling Point: 166-168°/15mm
Beilstein Reference: 2281302
MDL No.: MFCD00015126
Formula:
C₁₆H₂₀O₂Si Boiling Pt: 166…168 °C/15 mm Density: 1,033 Flash Pt: 175 °C(347 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015126 CAS Number: 2553-19-7 EINECS: 219-860-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...