2,2-Difluoroethanol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22201-03
B22201-06
CAAAB22201-03EA
99.22
CAD
CAAAB22201-03
CAAAB22201-06
2,2-Difluoroethanol 97%
2,2-Difluoroethanol
Purity: 97%
CAS No.: 359-13-7
Molecular Formula: HCF2CH2OH
Formula Weight: 82.05
Flash Point: 3°(37°F)
Refractive Index: 1.3335
Melting Point: -28°
Boiling Point: 95-96°
Beilstein Reference: 1731617
UN No.: 1987
MDL No.: MFCD00042248
Formula:
C₂H₄F₂O Boiling Pt: 95…96 °C Melting Pt: –28 °C Density: 1.296 Flash Pt: 3 °C (37 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042248 CAS Number: 359-13-7 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...