1-Aminocyclopentanecarboxylic acid 97+%

Supplier: Thermo Scientific Chemicals
Danger

B22179-03 B22179-14
CAAAAB22179-03EA 57.04 CAD
CAAAAB22179-03 CAAAAB22179-14
1-Aminocyclopentanecarboxylic acid 97+%
1-Aminocyclopentanecarboxylic acid

Purity: 97+% CAS No.: 52-52-8 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Melting Point: ca 320° dec. Beilstein Reference: 636626 Merck Reference: 14,2734 UN No.: 2811 MDL No.: MFCD00001381

Formula: C₆H₁₁NO₂
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: ∼ 320 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001381
CAS Number: 52-52-8
EINECS: 200-144-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,2734

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR