4-tert-Butylbenzenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-tert-Butylbenzene-1-sulfonyl chloride, 4-tert-Butylbenzene-1-sulphonyl chloride
B22174-09
B22174-18
CAAAB22174-09EA
71.1
CAD
CAAAB22174-09
CAAAB22174-18
4-tert-Butylbenzenesulfonyl chloride 98%
4-tert-Butylbenzenesulphonyl chloride
Purity: 98%
CAS No.: 15084-51-2
Molecular Formula: C10H13ClO2S
Formula Weight: 232.73
Odor: Irritating
Melting Point: 80-83°
Beilstein Reference: 1104346
UN No.: 3261
MDL No.: MFCD00007449
Formula:
C₁₀H₁₃ClO₂S MW: 232.73 g/mol Boiling Pt: 165 °C (18 -rr) Melting Pt: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007449 CAS Number: 15084-51-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...