Toluene-α-sulfonyl fluoride 99%

Supplier: Thermo Scientific Chemicals
Danger

B22146-03 B22146-06 B22146-14
CAAAB22146-03EA 48.03 CAD
CAAAB22146-03 CAAAB22146-06 CAAAB22146-14
Toluene-α-sulfonyl fluoride 99%
Toluene-α-sulphonyl fluoride

A powerful protease inhibitor


Purity: 99% CAS No.: 329-98-6 Molecular Formula: C7H7FO2S Formula Weight: 174.19 Melting Point: 92-94° Beilstein Reference: 2088311 UN No.: 2923 MDL No.: MFCD00007424

Formula: C₇H₇FO₂S
Boiling Pt: 80 - 85 °C (0.3 -rr)
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR