Diphenyl sulfoxide 98+%

Supplier: Thermo Scientific Chemicals

B22135-14 B22135-22 B22135-36
CAAAB22135-14EA 88.48 CAD
CAAAB22135-14 CAAAB22135-22 CAAAB22135-36
Diphenyl sulfoxide 98+%
Diphenyl sulphoxide

Purity: 98+% CAS No.: 945-51-7 Molecular Formula: C12H10OS Formula Weight: 202.28 Melting Point: 69-72° Boiling Point: 206-208°/13mm MDL No.: MFCD00002085

Formula: C₁₂H₁₀OS
Boiling Pt: 206…208 °C/13 mm
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002085
CAS Number: 945-51-7
EINECS: 213-415-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR