1-Methyl-2-phenylindole 99%
Supplier: Thermo Scientific Chemicals
B22105-09
B22105-18
B22105-30
CAAAB22105-09EA
49.61
CAD
CAAAB22105-09
CAAAB22105-18
CAAAB22105-30
1-Methyl-2-phenylindole 99%
1-Methyl-2-phenylindole
Purity: 99%
CAS No.: 3558-24-5
Molecular Formula: C15H13N
Formula Weight: 207.28
Melting Point: 98-99°
Beilstein Reference: 148770
Formula:
C₁₅H₁₃N Melting Pt: 98…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022892 CAS Number: 3558-24-5 EINECS: 222-618-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...