Nickel(II) chloride, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dichloride

B22085-14 B22085-22 B22085-36
CAAAAB22085-14EA 56.88 CAD
CAAAAB22085-14 CAAAAB22085-22 CAAAAB22085-36
Nickel(II) chloride, anhydrous 98%
Nickel (II) chloride

Purity: 98% CAS No.: 7718-54-9 Molecular Formula: Cl2Ni Formula Weight: 129.60 Odor: Odorless Melting Point: 1001° UN No.: 3288 MDL No.: MFCD00011142

Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1009
Density: 3.55
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 14,6505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR