Manganese(II) sulfate tetrahydrate 99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Manganese sulphate tetrahydrate, Manganese sulfate tetrahydrate, Manganous sulfate tetrahydrate, Manganous sulphate tetrahydrate
B22081-22
B22081-36
CAAAAB22081-22EA
61.46
CAD
CAAAAB22081-22
CAAAAB22081-36
Manganese(II) sulfate tetrahydrate 99% (metals basis)
Manganese (II) sulphate tetrahydrate
Purity: 99%
CAS No.: 10101-68-5
Molecular Formula: MnO4S.4H2O
Formula Weight: 223.06 (151.00anhy)
UN No.: 3077
MDL No.: MFCD00149160
Formula:
MnO₄S·4H₂O MW: 223.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149160 CAS Number: 10101-68-5 EINECS: 232-089-9 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...