Triethoxysilane 96%

Supplier: Thermo Scientific Chemicals
Danger

B22063-09 B22063-18 B22063-30
CAAAB22063-09EA 72.36 CAD
CAAAB22063-09 CAAAB22063-18 CAAAB22063-30
Triethoxysilane 96%
Triethoxysilane

Purity: 96% CAS No.: 998-30-1 Molecular Formula: C6H16O3Si Formula Weight: 164.28 Flash Point: 26°(78°F) Refractive Index: 1.3770 Melting Point: -170° Boiling Point: 134-135° Beilstein Reference: 1738989 UN No.: 3384 MDL No.: MFCD00009061

Formula: C₆H₁₆O₃Si
MW: 164.28 g/mol
Boiling Pt: 134…135 °C
Melting Pt: –170 °C
Density: 0.885
Flash Pt: 26 °C (78 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009061
CAS Number: 998-30-1
EINECS: 213-650-7
UN: 3489
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR