1,2,3-Trifluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22055-03
B22055-06
CAAAB22055-03EA
65.41
CAD
CAAAB22055-03
CAAAB22055-06
1,2,3-Trifluorobenzene 98+%
1,2,3-Trifluorobenzene
Purity: 98+%
CAS No.: 1489-53-8
Molecular Formula: C6H3F3
Formula Weight: 132.09
Flash Point: -3°(26°F)
Refractive Index: 1.4220
Boiling Point: 94-95°
Beilstein Reference: 1862388
UN No.: 2387
MDL No.: MFCD00012013
Formula:
C₆H₃F₃ Boiling Pt: 94…95 °C Density: 1.280 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00012013 CAS Number: 1489-53-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...