1-(4-Morpholino)cyclohexene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22004-14
B22004-22
CAAAB22004-14EA
50.24
CAD
CAAAB22004-14
CAAAB22004-22
1-(4-Morpholino)cyclohexene 97%
1-(4-Morpholino)cyclohexene
Purity: 97%
CAS No.: 670-80-4
Molecular Formula: C10H17NO
Formula Weight: 167.25
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.5146
Boiling Point: 118-120°/10mm
Beilstein Reference: 118696
MDL No.: MFCD00006163
Formula:
C₁₀H₁₇NO Boiling Pt: 118…120 °C/10 mm Density: 1.020 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006163 CAS Number: 670-80-4 EINECS: 211-579-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...