2-Methyl-1-butanethiol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylbutyl mercaptan, 1-Mercapto-2-methylbutane
B21949-06
B21949-14
CAAAB21949-06EA
88.64
CAD
CAAAB21949-06
CAAAB21949-14
2-Methyl-1-butanethiol 99%
2-Methyl-1-butanethiol
Purity: 99%
CAS No.: 1878-18-8
Molecular Formula: C5H12S
Formula Weight: 104.22
Flash Point: 19°(67°F)
Refractive Index: 1.4480
Boiling Point: 116-117°
UN No.: 1993
MDL No.: MFCD00004883
Formula:
C₅H₁₂S MW: 104.22 g/mol Boiling Pt: 116…117 °C Density: 0.845 Flash Pt: 19 °C (67 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004883 CAS Number: 1878-18-8 EINECS: 217-515-3 UN: 1111 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...