Phenol red sodium salt
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21948-06
B21948-14
CAAAAB21948-06EA
42.82
CAD
CAAAAB21948-06
CAAAAB21948-14
Phenol red sodium salt
Phenol red sodium salt
CAS No.: 34487-61-1
Molecular Formula: C19H13NaO5S
Formula Weight: 376.37
Melting Point: 285° dec.
Beilstein Reference: 6260026
Merck Reference: 14,7247
MDL No.: MFCD00066901
Formula:
C₁₉H₁₃NaO₅S Melting Pt: 285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00066901 CAS Number: 34487-61-1 EINECS: 252-057-8 Merck Index: 14,7247 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...