4'-Hydroxybiphenyl-4-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

B21946-09 B21946-88
CAAAB21946-09EA 342.86 CAD
CAAAB21946-09 CAAAB21946-88
4'-Hydroxybiphenyl-4-carboxylic acid 99%
4'-Hydroxybiphenyl-4-carboxylic acid

Purity: 99% CAS No.: 58574-03-1 Molecular Formula: C13H10O3 Formula Weight: 214.22 Melting Point: 292-295° MDL No.: MFCD00059078

Formula: C₁₃H₁₀O₃
Melting Pt: 292…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00059078
CAS Number: 58574-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR