4'-Hexylbiphenyl-4-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

B21900-03 B21900-06 B21900-14
CAAAB21900-03EA 67.31 CAD
CAAAB21900-03 CAAAB21900-06 CAAAB21900-14
4'-Hexylbiphenyl-4-carboxylic acid 99%
4'-Hexylbiphenyl-4-carboxylic acid

Purity: 99% CAS No.: 59662-48-5 Molecular Formula: C19H22O2 Formula Weight: 282.38 Melting Point: 222-224° MDL No.: MFCD00143232

Formula: C₁₉H₂₂O₂
Melting Pt: 222…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00143232
CAS Number: 59662-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR