(R)-(+)-1,2-Epoxypropane 99%

Supplier: Thermo Scientific Chemicals
Danger

B21884-06 B21884-14 B21884-22
CAAAB21884-06EA 135.56 CAD
CAAAB21884-06 CAAAB21884-14 CAAAB21884-22
(R)-(+)-1,2-Epoxypropane 99%
(R)-(+)-1,2-Epoxypropane

Purity: 99% CAS No.: 15448-47-2 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -37°(-35°F) Odor: Ether-like Boiling Point: 34-35° Beilstein Reference: 79764 Merck Reference: 14,7856 UN No.: 1280 MDL No.: MFCD00066211

Formula: C₃H₆O
Boiling Pt: 34…35 °C
Density: 0.829
Flash Pt: –37 °C (–35 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066211
CAS Number: 15448-47-2
UN: 1280
ADR: 3,I
Merck Index: 14,7856

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR