Indole-5-carbonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21810-03
B21810-06
CAAAB21810-03EA
83.58
CAD
CAAAB21810-03
CAAAB21810-06
CAAAB21810-14
Indole-5-carbonitrile 98+%
Indole-5-carbonitrile
Purity: 98+%
CAS No.: 15861-24-2
Molecular Formula: C9H6N2
Formula Weight: 142.16
Odor: Odorless
Melting Point: 104-108°
Beilstein Reference: 116738
MDL No.: MFCD00005669
Formula:
C₉H₆N₂ Melting Pt: 104…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005669 CAS Number: 15861-24-2 EINECS: 239-986-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...