6-Bromo-1-hexanol 97%

Supplier: Thermo Scientific Chemicals
Warning

B21803-03 B21803-06 B21803-14
CAAAAB21803-03EA 61.3 CAD
CAAAAB21803-03 CAAAAB21803-06 CAAAAB21803-14
6-Bromo-1-hexanol 97%
6-Bromo-1-hexanol

Purity: 97% CAS No.: 4286-55-9 Molecular Formula: Br(CH2)6OH Formula Weight: 181.08 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4820 Boiling Point: 105-106°/5mm MDL No.: MFCD00002983

Formula: BrH₂C(CH₂)₄CH₂OH
Boiling Pt: 105…106 °C/5 mm
Density: 1,384
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002983
CAS Number: 4286-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR