Trifluoromethanesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21774-06
B21774-14
B21774-22
CAAAB21774-06EA
116.29
CAD
CAAAB21774-06
CAAAB21774-14
CAAAB21774-22
Trifluoromethanesulfonyl chloride 98%
Trifluoromethanesulphonyl chloride
Purity: 98%
CAS No.: 421-83-0
Molecular Formula: CF3SO2Cl
Formula Weight: 168.52
Storage Temperature: 2°C to 8°C
Refractive Index: 1.3340
Odor: Pungent
Boiling Point: 32°
Beilstein Reference: 1812016
UN No.: 3265
MDL No.: MFCD00007451
Formula:
CClF₃O₂S Boiling Pt: 32 °C Density: 1.593 Storage Temperature: Ambient |
MDL Number:
MFCD00007451 CAS Number: 421-83-0 EINECS: 207-009-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...