Dimethyl sulfone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl sulphone, Dimethylsulfone, Methyl sulfone, Dimethylsulphone
B21747-0B
B21747-30
CAAAAB21747-0BEA
289.14
CAD
CAAAAB21747-0B
CAAAAB21747-30
Dimethyl sulfone 99%
Dimethyl sulphone
Purity: 99%
CAS No.: 67-71-0
Molecular Formula: C2H6O2S
Formula Weight: 94.13
Flash Point: 143°(289°F)
Odor: Odorless
Boiling Point: 238°
Beilstein Reference: 1737717
Merck Reference: 14,3258
MDL No.: MFCD00007566
Formula:
C₂H₆O₂S MW: 94.13 g/mol Boiling Pt: 238 °C Melting Pt: 108…111 °C Density: 1.45 Flash Pt: 143 °C(289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007566 CAS Number: 67-71-0 EINECS: 200-665-9 Merck Index: 14,3258 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...