α-Bromo-o-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21717-14
B21717-22
CAAAB21717-14EA
103.49
CAD
CAAAB21717-14
CAAAB21717-22
α-Bromo-o-xylene 98%
α-Bromo-o-xylene
Purity: 98%
CAS No.: 89-92-9
Molecular Formula: C8H9Br
Formula Weight: 185.07
Storage Temperature: 2°C to 8°C
Flash Point: 82°(179°F)
Refractive Index: 1.5750
Melting Point: 18-20°
Boiling Point: 216-217°
Beilstein Reference: 636333
Merck Reference: 14,10090
UN No.: 1701
MDL No.: MFCD00000174
Formula:
C₈H₉Br Boiling Pt: 216…217 °C Melting Pt: 18…20 °C Density: 1.381 Flash Pt: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000174 CAS Number: 89-92-9 EINECS: 201-951-6 UN: 1701 ADR: 6.1,II Merck Index: 14,10090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...