4-Bromo-2-methylanisole 98+%

Supplier: Thermo Scientific Chemicals

B21621-09 B21621-18
CAAAAB21621-09EA 53.4 CAD
CAAAAB21621-09 CAAAAB21621-18
4-Bromo-2-methylanisole 98+%
4-Bromo-2-methylanisole

Purity: 98+% CAS No.: 14804-31-0 Molecular Formula: C8H9BrO Formula Weight: 201.06 Melting Point: 66-69° Boiling Point: 124-125°/25mm MDL No.: MFCD01321139

Formula: C₈H₉BrO
Boiling Pt: 124…125 °C/25 mm
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01321139
CAS Number: 14804-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR