2-Chlorostyrene 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0.1%
B21551-06
B21551-14
CAAAAB21551-06EA
141.88
CAD
CAAAAB21551-06
CAAAAB21551-14
CAAAAB21551-22
2-Chlorostyrene 97% stabilized
2-Chlorostyrene
Purity: 98%
CAS No.: 2039-87-4
Molecular Formula: C8H7Cl
Formula Weight: 138.60
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.5650
Odor: Pungent
Melting Point: -63°
Boiling Point: 189°
Beilstein Reference: 2038863
MDL No.: MFCD00000567
Formula:
C₈H₇Cl Boiling Pt: 189 °C Melting Pt: –63 °C Density: 1.091 Flash Pt: 65 °C (149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000567 CAS Number: 2039-87-4 EINECS: 218-026-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...