1,4-Dichlorobutane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21337-0B
B21337-30
CAAAAB21337-0BEA
165.9
CAD
CAAAAB21337-0B
CAAAAB21337-30
1,4-Dichlorobutane 97%
1,4-Dichlorobutane
Purity: 97%
CAS No.: 110-56-5
Molecular Formula: Cl(CH2)4Cl
Formula Weight: 127.01
Flash Point: 40°(104°F)
Refractive Index: 1.4540
Odor: Aromatic
Melting Point: -38°
Boiling Point: 155°
Beilstein Reference: 635673
UN No.: 1993
MDL No.: MFCD00001011
Formula:
ClH₂C(CH₂)₂CH₂Cl Boiling Pt: 155 °C Melting Pt: –38 °C Density: 1.139 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001011 CAS Number: 110-56-5 EINECS: 203-778-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...