2-Methylhydroquinone 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methylhydroquinone
B21241-0B
B21241-30
CAAAAB21241-0BEA
341.28
CAD
CAAAAB21241-0B
CAAAAB21241-30
2-Methylhydroquinone 99%
2-Methylhydroquinone
Purity: 99%
CAS No.: 95-71-6
Molecular Formula: C7H8O2
Formula Weight: 124.14
Flash Point: 172°(341°F)
Melting Point: 127-130°
Boiling Point: 284-286°
Beilstein Reference: 2041489
MDL No.: MFCD00002345
Formula:
C₇H₈O₂ MW: 124.14 g/mol Boiling Pt: 284…286 °C Melting Pt: 127…130 °C Flash Pt: 172 °C(341 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002345 CAS Number: 95-71-6 EINECS: 202-443-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...