D-Threonine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-D-Thr-OH, (2R,3S)-2-Amino-3-hydroxybutanoic acid
B21177-06
B21177-14
B21177-22
CAAAB21177-06EA
63.67
CAD
CAAAB21177-06
CAAAB21177-14
CAAAB21177-22
D-Threonine 99%
D-Threonine
Purity: 99%
CAS No.: 632-20-2
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Melting Point: ca 274° dec.
Beilstein Reference: 1721643
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: ∼ 274 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064269 CAS Number: 632-20-2 EINECS: 211-171-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...