Isoamyl butyrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopentyl butyrate, Isoamyl Butyrate

B21103-22 B21103-36
CAAAB21103-22EA 40.29 CAD
CAAAB21103-22 CAAAB21103-36
Isoamyl butyrate 98%
Isoamyl butyrate

Purity: 98% CAS No.: 106-27-4 Molecular Formula: CH3CH2CH2CO2CH2CH2CH(CH3)2 Formula Weight: 158.24 Flash Point: 59°(138°F) Refractive Index: 1.411 Odor: Fruit-like Melting Point: -73° Boiling Point: 178° Merck Reference: 14,5115 UN No.: 2620 MDL No.: MFCD00044888

Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 178 °C
Melting Pt: –73 °C
Density: 0.88
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044888
CAS Number: 106-27-4
EINECS: 203-380-8
UN: 2620
ADR: "3",III
Merck Index: 14,5115

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR