Sodium dehydracetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dehydracetic acid sodium salt, Dehydroacetic acid sodium salt, Sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide, Sodium dehydroacetate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate

B21060-22 B21060-36
CAAAB21060-0EEA 1678.91 CAD
CAAAB21060-0E CAAAB21060-22 CAAAB21060-36
Sodium dehydracetate 97%
Sodium dehydracetate

Purity: 97% CAS No.: 4418-26-2 Molecular Formula: C8H7NaO4 Formula Weight: 190.13 pH: 8.8-9.3 Melting Point: ca 295° dec. MDL No.: MFCD00040583

Formula: C₈H₇NaO₄
MW: 190.13 g/mol
Melting Pt: ∼ 295 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040583
CAS Number: 4418-26-2
EINECS: 224-580-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR