Methyl-2-amino-4,5-dimethoxybenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4,5-dimethoxyanthranilate, 2-Amino-4,5-dimethoxy benzoate, Methyl 2-amino-4,5-dimethoxybenzoate, 2-Amino-4,5-dimethoxybenzoic acid methyl ester
B21002-09
B21002-18
CAAAB21002-09EA
33.65
CAD
CAAAB21002-09
CAAAB21002-18
Methyl-2-amino-4,5-dimethoxybenzoate 98%
Methyl 2-amino-4,5-dimethoxybenzoate
Purity: 98%
CAS No.: 26759-46-6
Molecular Formula: C10H13NO4
Formula Weight: 211.22
Melting Point: 128-130°
Beilstein Reference: 2724754
Formula:
C₁₀H₁₃NO₄ Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014904 CAS Number: 26759-46-6 EINECS: 247-967-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...