1H,1H,2H,2H-Perfluoro-1-iodooctane 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
B20984-06
B20984-14
CAAAAB20984-06EA
62.41
CAD
CAAAAB20984-06
CAAAAB20984-14
1H,1H,2H,2H-Perfluoro-1-iodooctane 97% stabilized
1H,1H,2H,2H-Perfluoro-1-iodooctane
Purity: 97%
CAS No.: 2043-57-4
Molecular Formula: CF3(CF2)5CH2CH2I
Formula Weight: 474.00
Flash Point: 150°(302°F)
Refractive Index: 1.3600
Melting Point: ca 20°
Boiling Point: 178-180°
Beilstein Reference: 1887105
MDL No.: MFCD00039410
Formula:
C₈H₄F₁₃I Boiling Pt: 178…180 °C Melting Pt: ∼ 20 °C Density: 1.940 Flash Pt: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039410 CAS Number: 2043-57-4 EINECS: 218-056-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...